Stampa…

Benactyzine hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Pericolo

B0053-5G
TCIAB0053-5GEA 105 EUR
TCIAB0053-5G
Benactyzine hydrochloride ≥98.0% (by HPLC, titration analysis)
Benactyzine hydrochloride
Formula: C₂₀H₂₅NO₃
MDL Number: MFCD00012624
CAS Number: 57-37-4
EINECS: 200-324-4
UN: 2811
ADR: 6.1,III
Merck Index: 14,01029

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 175.0 to 179.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR