Stampa…

Argatroban monohydrate ≥99.0% (by HPLC)

Fornitore: TCI

A2705-25MG
TCIAA2705-25MGEA 135 EUR
TCIAA2705-25MG
Argatroban monohydrate ≥99.0% (by HPLC)
Argatroban monohydrate
Formula: C₂₃H₃₆N₆O₅S·H₂O
Punto di fusione: 180°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00895735
CAS Number: 141396-28-3
Merck Index: 14,00779

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Elemental analysis (Nitrogen) 15.50 to 16.50 %
Specific rotation [a]20/D +160.0 to +170.0 deg (C=0.8, MeOH) (calcd.on anh.substance)
Water 2.5 to 4.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR