Stampa…

α-Acetyl-γ-butyrolactone ≥99.0% (by GC)

Fornitore: TCI
Attenzione

A0681-25G A0681-500G
TCIAA0681-25GEA 16 EUR
TCIAA0681-25G TCIAA0681-500G
α-Acetyl-γ-butyrolactone ≥99.0% (by GC)
α-Acetyl-γ-butyrolactone
Formula: C₆H₈O₃
Flash Pt: 135°C
MDL Number: MFCD00005394
CAS Number: 517-23-7
EINECS: 208-235-2
Merck Index: 14,00083
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebb790caba09966fa0188202a243d5a4fdff72c3&vid=ebb790caba09966fa0188202a243d5a4fdff72c3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR