Stampa…
Amberlite® XAD®-2, Pore size: 90 Å mean 20 - 60 mesh ion-exchange resin, Supelco®
Fornitore: Merck
M-Clarity™ quality level = MQ100
SU853005
SU853005EA 1260 EUR
SU853005
Amberlite® XAD®-2, Pore size: 90 Å mean 20 - 60 mesh ion-exchange resin, Supelco®
Amberlite® XAD®-2
Amberlite® XAD®-2 is a strongly acidic ion-exchange resin.
  • Surface area: ~300 m²/g
Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.
Amberlite® XAD®-2 may be used as a stationary phase material for the solid phase extraction (SPE) of phenolic acids and flavonols from honey samples prior to their identification by high performance liquid chromatography-diode array detection (HPLC-DAD),[1] phthalate esters (PAEs) from hydroalcoholic food beverages prior to their determination by gas chromatography (GC) equipped with flame ionization detector (FID) as well as ion-trap mass spectrometry detector (IT/MS), benzoic and sorbic acids in quince jam prior to their analysis by isocratic HPLC combined with photodiode array (PDA) detector and organophosphorus pesticides in natural and drinking waters prior to their determination by GC-FID or GC coupled to nitrogen-phosphorus detector (NPD).
CAS Number: 9003-70-7