Stampa…
Cole-Parmer® Stuart SI-200 Microplate Shaking Incubators
Fornitore: Cole Parmer
79520-10 79520-12
STUA79520-10EA 4790 EUR
STUA79520-10 STUA79520-12
Cole-Parmer® Stuart SI-200 Microplate Shaking Incubators
Agitatori Agitatori per micropiastre, accessori
Specifically designed to mix small volumes of liquid in a temperature-controlled environment.
  • Microprocessor controlled shaking speed ensures accuracy
  • High shaking speed up to 1250 min⁻¹ (rpm)
  • 1,5 mm diameter orbit is ideal for microplates and microtubes
  • Easily set and view temperature and speed on the digital display
  • Unique retractable platform offers easy loading and unloading
These microplate shaking incubators combine high speed with tiny orbit to create the vibrational shaking action required for small sample volumes. All models feature a timerMore Product Information