Stampa…
Cole-Parmer® Intelli-Mixer RM-2 Rotator, Vortex Shaker
Fornitore: Cole Parmer
51202-20
STUA51202-20EA 1890 EUR
STUA51202-20
Cole-Parmer® Intelli-Mixer RM-2 Rotator, Vortex Shaker
Agitatori Agitatori vortex
Rotator, vortex mixer, blotter all in one.
  • 34 modes of operation
  • 360° rotation
  • Accepts 0,5 to 50 ml tubes
  • Cold room safe
Powered by V-Spin, the Intelli-Mixer RM-2 is the latest in cutting-edge science. V-spin harnesses the energy of a stepper motor, allowing for a wide variety of mixing, vibrating, and rotating cycles. Featuring 15 Mix, 12 rocking, 4 blotting, and 3 custom modes all in one versitile unit.
Perform a variety of mixing in tubes 0,5 to 50 ml or simultaneously vortex up to 42 eppendorf tubes with 2 different vortex frequencies. EnsureMore Product Information
Visualizza i codici
Specifiche
Environmentally Preferable
Velocità 1 - 99 min⁻¹
L×P×A 406,4×171,5×133 mm
Elettrico 120/230 VAC