Stampa…
Undergrad hotplates, Stuart®
Fornitore: Cole Parmer
04807-50 04807-52
STUA04807-50EA 723 EUR
STUA04807-50 STUA04807-52
Undergrad hotplates, Stuart®
Termostati Piastre riscaldanti
Get accurate surface temperature in real time.
  • Glazed with BioCote® for antimicrobial protection
  • Compact, stackable design takes up less benchtop and storage space
  • Hot warning light remains on even when unplugged
  • Built with tough aluminium casing for longevity
  • Designed to prevent accidental spillage on control panel
  • IP 32 rated for water and dust resistance
Designed with microprocessor-controlled dual thermocouples, these hotplates offer accurate temperature control and protection from overheating. A coloured LED indicator surrounds the heat controlMore Product Information