Stampa…

Anti-Acetylated Lysine Rabbit Polyclonal Antibody

Fornitore: Merck Millipore (Oncogene)
ST1027-50
ST1027-50EA 680 EUR
ST1027-50
Anti-Acetylated Lysine Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: Acetylated Lysine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Pan
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR