Stampa…
Hollow fibre filter modules, PS membrane, MidiKros®
Fornitore: Spectrum Laboratories
D02-S05U-05-N D04-S05U-05-N D04-S050-05-S D04-S050-05-P D04-S050-05-N D04-S010-05-S D04-S010-05-P D04-S500-05-N D04-S010-05-N D04-S05U-05-S D04-S500-05-P D02-S500-05-S D02-S500-05-P D02-S500-05-N D06-S05U-05-N D06-S050-05-S D06-S050-05-P D06-S050-05-N D06-S010-05-S D06-S010-05-P D06-S500-05-N D06-S010-05-N D02-S05U-05-P D06-S05U-05-P D06-S05U-05-S D06-S500-05-S D06-S500-05-P D02-S050-05-S D02-S050-05-P D02-S050-05-N D02-S010-05-S D02-S010-05-P D02-S010-05-N D04-S500-05-S D02-S05U-05-S
SPECD02-S05U-05-NEA 544 EUR
SPECD02-S05U-05-N SPECD04-S05U-05-N SPECD04-S050-05-S SPECD04-S050-05-P SPECD04-S050-05-N SPECD04-S010-05-S SPECD04-S010-05-P SPECD04-S500-05-N SPECD04-S010-05-N SPECD04-S05U-05-S SPECD04-S500-05-P SPECD02-S500-05-S SPECD02-S500-05-P SPECD02-S500-05-N SPECD06-S05U-05-N SPECD06-S050-05-S SPECD06-S050-05-P SPECD06-S050-05-N SPECD06-S010-05-S SPECD06-S010-05-P SPECD06-S500-05-N SPECD06-S010-05-N SPECD02-S05U-05-P SPECD06-S05U-05-P SPECD06-S05U-05-S SPECD06-S500-05-S SPECD06-S500-05-P SPECD02-S050-05-S SPECD02-S050-05-P SPECD02-S050-05-N SPECD02-S010-05-S SPECD02-S010-05-P SPECD02-S010-05-N SPECD04-S500-05-S SPECD02-S05U-05-S
Hollow fibre filter modules, PS membrane, MidiKros®
Filtri Sistemi a flusso tangenziale
Hollow fibre filter modules are particularly useful in tangential flow filtration because processes can easily be scaled-up from small volume applications to full-scale production.
  • Hollow fibre membranes are anisotropic in structure
  • The filter housings are engineered to handle higher pressures and autoclaving
  • Low protein binding increases the product yield of each processing run
  • These filters have FLL/FLL connections and come in three effective lengths
Tangential flow filtration is used in both microfiltration and ultra-filtration applications. Ultra-More Product Information
Visualizza i codici
Specifiche
Environmentally Preferable
Raccordo di ingresso/uscita connessioni Female Luer lock/female Luer lock