Stampa…

Zinc stearate, powder USP

Fornitore: Spectrum Chemical

Synonyms: Stearic acid zinc salt, Zinc(II) stearate

ZI115-500GM ZI115-12KG ZI115-2.5KG
SPCMZI115-500GMEA 142 EUR
SPCMZI115-500GM SPCMZI115-12KG SPCMZI115-2.5KG
Zinc stearate, powder USP
Zinco stearato

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₃₆H₇₀O₄Zn
Peso molecolare: 632.34 g/mol
Punto di fusione: 140
Flash Pt: 276
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00013031
CAS Number: 557-05-1
Merck Index: 13,10212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8ad7f3e101681798edbd54e57ad5d2ac82b2e5d&vid=435308cdcb35862fe6cc06384da2ef10d84c5700&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Assay (ZnO) 12.5 - 14.0%
Alkalies and Alkaline Earths max. 1.0 %
Arsenic max. 1.5 ppm
Lead max. 10 ppm
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR