Stampa…

Sodium stearyl fumarate 99.0-101.5% (by anhydrous basis) NF

Fornitore: Spectrum Chemical

S1448-25KGBL S1448-500GM S1448-25GM
SPCMS1448-25KGBLEA 44720 EUR
SPCMS1448-25KGBL SPCMS1448-500GM SPCMS1448-25GM
Sodium stearyl fumarate 99.0-101.5% (by anhydrous basis) NF
Sodium stearyl fumarate

Sodium stearyl fumarate, NF is a water-soluble tablet lubicant.


Manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₂₂H₄₀O₄
MDL Number: MFCD00236640
CAS Number: 4070-80-8

Visualizza i codici

Risultati del test di specificazione

Assay (Anhydrous Basis) 99.0 - 101.5%
Saponification Value 142.2 - 146.0
Water Determination Max. 5.0%
Lead (Pb) Max. 10 ppm
Heavy Metals Max. 20 ppm
Sodium Stearyl Maleate And Stearyl Alcohol To Pass Test
Residual Solvents To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR