Stampa…

Polysorbate 60 NF

Fornitore: Spectrum Chemical

PO136-200LTBL PO136-20LT PO136-4LTGL
SPCMPO136-200LTBLEA 8100 EUR
SPCMPO136-200LTBL SPCMPO136-20LT SPCMPO136-4LTGL
Polysorbate 60 NF
Polysorbate 60
Formula: C₆₄H₁₂₆O₂₆
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01780110
CAS Number: 9005-67-8
EINECS: 500-020-4

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR