Stampa…

Poloxamer 188 NF

Fornitore: Spectrum Chemical

P1169-12KG P1169-2.5KG P1169-500GM
SPCMP1169-12KGEA 2590 EUR
SPCMP1169-12KG SPCMP1169-2.5KG SPCMP1169-500GM
Poloxamer 188 NF
Poloxamer 188 (Lutrol® F68, Pluronic® F-68, Synperonic® F68)

Poloxamer 188, NF is a useful surfactant for the formulation of cosmetics and pharmaceuticals because it increases miscibility. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a
MDL Number: MFCD00082049
CAS Number: 9003-11-6

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR