Stampa…

Acido nicotinico (Niacin) 98.0-102.0% (dried basis), in polvere USP

Fornitore: Spectrum Chemical
Attenzione

NI100-100GM NI100-12KGBL NI100-1KG
SPCMNI100-100GMEA 200 EUR
SPCMNI100-100GM SPCMNI100-12KGBL SPCMNI100-1KG
Acido nicotinico (Niacin) 98.0-102.0% (dried basis), in polvere USP
Acido nicotinico (Niacin)

Niacin, is also referred to as vitamin B3 and nicotinic acid and is one of the essential human nutrients. It is a colorless, water soluble derivative of pyridine. As a food supplement it is available over the counter but can also be found naturally in various food groups such as meat, fruit and vegetables and seeds.


Certificazioni: Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₆H₅NO₂
MDL Number: MFCD00006391
CAS Number: 59-67-6

Visualizza i codici

Risultati del test di specificazione

Assay (Dried Basis) 98.0 - 102.0 %
Loss On Drying max.1.0 %
Residue On Ignition max. 0.1 %
Chloride (Cl) max. 0.02 %
Sulfates (SO₄) max. 0.02 %
Cadmium As reported
Lead (Pb) As reported
ARSENIC (As) As reported
Mercury (Hg) As reported
Related compounds To pass test
Identification To pass test
Certified Kosher
Certified Halal
Residual solvents To pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR