Stampa…

Metile-4-idrossibenzoato ≥98.0% NF

Fornitore: Spectrum Chemical

Synonyms: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate

ME163-500GM ME163-50KGBL
SPCMME163-500GMEA 185 EUR
SPCMME163-500GM SPCMME163-50KGBL
Metile-4-idrossibenzoato ≥98.0% NF
Metile-4-idrossibenzoato

Methylparaben, NF is an excipient used as a preservative. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark.

Formula: HOC₆H₄CO₂CH₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002352
CAS Number: 99-76-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84321052052f4be2a53d8d7ebac7fabe6fc5a435&vid=ede765fad275b2515a8c57ba4499bff313a18983&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Assay (C₈H₈O₃) 98.0 - 102.0 %
Color of Solution To Pass Test
Acidity To Pass Test
Residue On Ignition max. 0.1 %
Related Substances To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR