Stampa…

Erythrosin B disodium salt

Fornitore: Spectrum Chemical
Attenzione

Synonyms: Erythrosina 239, C.I. 45430

FD135-100GM FD135-25GM FD135-500GM
SPCMFD135-100GMEA 196 EUR
SPCMFD135-100GM SPCMFD135-25GM SPCMFD135-500GM
Erythrosin B disodium salt
Erythrosin B disodium salt

FD and C Red No. 3 | Non-graded FD and C Red No. 3 supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₀H₆I₄Na₂O₅
Peso molecolare: 879.86 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00144257
CAS Number: 16423-68-0
EINECS: 240-474-8

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR