Stampa…

2-(Dimethylamino)-ethanol 99

Fornitore: Spectrum Chemical

Synonyms: DMAE

D2399-2.5LTGL D2399-200LTBL D2399-20LT
SPCMD2399-2.5LTGLEA 438 EUR
SPCMD2399-2.5LTGL SPCMD2399-200LTBL SPCMD2399-20LT
2-(Dimethylamino)-ethanol 99
2-Dimetilaminoetanolo

N,N-Dimethylethanolamine is a transparent, pale yellow liquid and primary alcohol that is used as a building block for the synthesis of cationic flocculants and ion exchange resins. It is also used used as a chemical intermediate for pharmaceuticals, dyes, corrosion inhibitors, and emulsifiers and as an additive to boiler water, paint removers, and amino resins.


GHS hazard : Corrosion;Flame;Skull and crossbones;Exclamation mark
UN2051

Formula: C₄H₁₁NO
Peso molecolare: 89.14 g/mol
Punto di fusione: -59
Densità: 0,887
Flash Pt: 39
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002846
CAS Number: 108-01-0
UN: 2051
ADR: 8.3 II
Merck Index: 13,02863

Visualizza i codici

Risultati del test di specificazione

Assay 0.99
Color (APHA) max. 20
Water max. 0.1 %
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR