Stampa…

L(+)-Cisteina cloridrata monoidrata USP

Fornitore: Spectrum Chemical

Synonyms: (R)-(+)-Cysteine hydrochloride monohydrate, (R)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate, L-Cysteine HCI Monohydrate, H-Cys-OH.HCl.H2O, Cysteine Hydrochloride, Monohydrate, Cysteine Hydrochloride monohydrate

C1473-12KGBL C1473-1KG C1473-50KGBL
SPCMC1473-12KGBLEA 0 EUR
SPCMC1473-12KGBL SPCMC1473-1KG SPCMC1473-50KGBL
L(+)-Cisteina cloridrata monoidrata USP
L(+)-Cisteina cloridrata monoidrata

BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Formula: HSCH₂CH(NH₂)COOH·HCl·H₂O
Peso molecolare: 175.64 g/mol
Punto di fusione: 168…170 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00065606
CAS Number: 7048-04-6
EINECS: 200-157-7
Merck Index: 13,02810
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18703c57a6b5f299a5f38f5d92b333d3d65de497&vid=10395059845247050509d84df67aefcb3c98806c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Assay 98.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR