Stampa…

tri-Calcio citrato tetraidrato, powder USP

Fornitore: Spectrum Chemical

Synonyms: tri-Calcium dicitrate tetrahydrate, Calcium citrate tribasic tetrahydrate

C1102-125LBBL C1102-12KG C1102-2.5KG
SPCMC1102-125LBBLEA 3300 EUR
SPCMC1102-125LBBL SPCMC1102-12KG SPCMC1102-2.5KG
tri-Calcio citrato tetraidrato, powder USP
tri-Calcio citrato tetraidrato

Form: Powder

Formula: C₁₂H₁₀Ca₃O₁₄·4H₂O
Peso molecolare: 570.5 g/mol
MDL Number: MFCD00150786
CAS Number: 5785-44-4
EINECS: 212-391-7

Visualizza i codici

Risultati del test di specificazione

Assay 97.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR