Stampa…

Aluminum monostearate NF

Fornitore: Spectrum Chemical

Synonyms: Dihydroxy(stearoyloxy)aluminum, Aluminum monostearate technical(Hard and soft aluminium mixture), Aluminum dihydroxide stearate

AL228-12KGBL AL228-2.5KGBL AL228-50KGBL
SPCMAL228-12KGBLEA 0 EUR
SPCMAL228-12KGBL SPCMAL228-2.5KGBL SPCMAL228-50KGBL
Aluminum monostearate NF
Aluminum monostearate

Aluminum Monostearate, NF is also known as Dihydroxyaluminum stearate.


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

Formula: C₁₈H₃₇AlO₄
Peso molecolare: 344.4655 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00019932
CAS Number: 7047-84-9

Visualizza i codici

Risultati del test di specificazione

Assay (Al₂O₃) 14.5-16.5%
Loss On Drying max.2.0 %
Arsenic (As) max. 4 ppm
Heavy Metals max. 50 Ug/G
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR