Stampa…

Adenine hydrochloride

Fornitore: Spectrum Chemical

Synonyms: 7H-Purin-6-amine hydrochloride, Purin-6-amine hydrochloride, 6-Aminopurine hydrochloride

A3192-1KGBL A3192-25KGBL
SPCMA3192-1KGBLEA 4800 EUR
SPCMA3192-1KGBL SPCMA3192-25KGBL
Adenine hydrochloride
Adenine hydrochloride
Formula: C₅H₅N₅·HCl
Peso molecolare: 171.59 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00150864
CAS Number: 2922-28-3

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR