Stampa…
A
Fornitore: Spectrum Chemical
Synonyms: Anti-foaming agent
A1302-200LTBL A1302-20LTBL A1302-4LTPL A1302-100ML A1302-500MLPL
SPCMA1302-200LTBLEA 0 EUR
SPCMA1302-200LTBL SPCMA1302-20LTBL SPCMA1302-4LTPL SPCMA1302-100ML SPCMA1302-500MLPL
A
Silicone antischiuma
Antifoam A is also known as polydimethylsiloxane and is part of a group of polymeric organosilicon compounds known as silicones.
It is a chemical additive that reduces and hinders the formation of foam in industrial process liquids. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.