Stampa…
PCR microtubes without caps, 0,2 ml, Amplitube™
Fornitore: Simport Scientific
T325-12B T325-12G T325-12N T325-12R T325-12Y
SIMPT325-12BEA 53.1 EUR
SIMPT325-12B SIMPT325-12G SIMPT325-12N SIMPT325-12R SIMPT325-12Y
PCR microtubes without caps, 0,2 ml, Amplitube™
Tubi Tubi per PCR
Made of PP, translucent.
  • Choice of colorless and four non-cytotoxic and non-metallic colors
  • Very good chemical resistance
  • Very good temperature resistance
This thin wall 0,2 ml tube is very useful when processing smaller volumes. It offers optimum contact with thermal cycler blocks. The ultrathin wall will ensure rapid thermal transfer and a significant reduction in cycle and PCR reaction time. Specially designed with a highly polished surface and a round bottom for maximum sample recovery.
Informazioni per l'ordine: Caps must be ordered seperately.More Product Information