Stampa…
Dissection boards, disposable, DispoCut™
Fornitore: Simport Scientific
M630-1 M630-2 M630-3
SIMPM630-1EA 518 EUR
SIMPM630-1 SIMPM630-2 SIMPM630-3
Dissection boards, disposable, DispoCut™
Tavole per dissezione
These dissection boards are made from a copolymer resin in order to increase impact and temperature performance. The board is strong yet inexpensive enough to throw away. It is especially developed to provide a clean, safer and more efficient way of handling infectious tissue specimens for the pathologist.
  • Can be used on both sides
  • Printed with helpful imperial and metric dimensional scales in inches, centimetres and millimetres
  • Chemically inert, withstands most oils, solvents and water (at regular temperatures)
  • Surface will not dull knifes
  • Perfect for use