Stampa…
Weighing dishes, anti-static
Fornitore: Simport Scientific
D250-3 D250-1 D250-2
SIMPD250-3EA 131 EUR
SIMPD250-3 SIMPD250-1 SIMPD250-2
Weighing dishes, anti-static
Navicelle Navicelle per pesata
These weighing dishes are safe, contaminant-free, biologically inert economical containers for weighing liquid or powdered samples in the laboratory.
  • Resistant to diluted acids, aqueous solutions, alcohols and bases
  • Flat bottom ensures perfect stability on countertops
  • Smooth surface providing accurate pour-outs with minimal sample loss
  • Facilitate weighing of static-affected samples
  • Ideal for many applications such as weighing, dispensing or storing
Dishes can also be used as quick freeze trays for sample material, discard trays for broken ampules, orMore Product Information