Stampa…
Test charts
Fornitore: Industrial Physics
VF2325 VF2347 VF2346 VF2345 VF2344 VF2321 VF2317 VF2343 VF3005 VF3004 VF3003 VF3007 VF3006 VF2319 VF3008 VF2354 VF3000 VF3001 VF3002 VF2323 VF2316
SIMEVF2325EA 262.44 EUR
SIMEVF2325 SIMEVF2347 SIMEVF2346 SIMEVF2345 SIMEVF2344 SIMEVF2321 SIMEVF2317 SIMEVF2343 SIMEVF3005 SIMEVF3004 SIMEVF3003 SIMEVF3007 SIMEVF3006 SIMEVF2319 SIMEVF3008 SIMEVF2354 SIMEVF3000 SIMEVF3001 SIMEVF3002 SIMEVF2323 SIMEVF2316
Test charts
Grafici
Test charts are used for testing physical properties of coating, lacquers, and inks.
  • Dimensions from A6 up to A4
  • Film laminated for an excellent solvent, chemical resistance, and film spread
  • Section for the date, time, and test number
Suitable for determining hiding power, opacity, and spreading rate in coating laboratories and paint production.
Informazioni per l'ordine: A certificate is available for charts with an optical brightener.