Stampa…
Crimpers and Decappers
Fornitore: Trajan Scientific and Medical
308990 311990 311991 313990 313991 313992 320990 320991 320992
SGEA308990EA 285 EUR
SGEA308990 SGEA311990 SGEA311991 SGEA313990 SGEA313991 SGEA313992 SGEA320990 SGEA320991 SGEA320992
Crimpers and Decappers
Capsulatori e decapsulatori
Crimping tools have advanced the standards of manual crimpers and decappers by addressing all of the deficiencies associated with traditional blocky, metal tools.
  • Reliabily seal crimps caps manually with less physical strain
  • constructed of tough, light weight plastic that is incredibly durable but weighs much less than metal crimping tools
  • Curved handles create a natural grip, greatly improving comfort over metal grips
  • Unique bottom pull technology allows for steadier operation in comparison to top-push handle designs
Application: Low volume crimping/More Product Information