Stampa…
Siringhe a tenuta di gas da 1 a 100 ml
Fornitore: Trajan Scientific and Medical
008020 008025 008100 008102 008420 008425 008500 008502 008700 008760 008762 008900 008960 008962 009462 009463 009660 009760 008102CAL 008100CAL 008500CAL 008025CAL 008962CAL
549-0332EA 95 EUR
549-0332 549-0333 549-0334 548-0948 549-0335 549-0336 549-0337 548-0949 549-0338 549-0339 548-0950 549-0340 549-0341 548-0951 549-0342 548-0952 549-0343 549-0344 SGEA008102CAL SGEA008100CAL SGEA008500CAL SGEA008025CAL SGEA008962CAL
Siringhe a tenuta di gas da 1 a 100 ml
Siringhe Siringhe per cromatografia
Realizzate in vetro borosilicato, puntali dello stantuffo in PTFE e coni Luer Kel-F® o in PTFE dove applicabile.
  • Assembled with an interference fit between plunger tip and barrel to effect the gas sealing properties of the syringe
  • Plungers are replaceable on these syringes
  • Suitable for manual and dispenser/diluter application
Diametro esterno del cilindro: 1 ml=9 mm; 2,5 ml=11 mm; 5 ml=14 mm; 10 ml=18 mm; 25 ml=27 mm; 50 ml=32,8 mm; 100 ml=40,8 mm