Stampa…
Thermosept® NKZ
Fornitore: SCHUELKE MAYR
146503
SCUE146503EA 0 EUR
SCUE146503
Thermosept® NKZ
Soluzioni detergenti Disinfettanti
Neutraliser based on citric acid for the automated reprocessing of medical devices and laboratory accessories.
  • Free of phosphates and surfactants
  • Gentle on material
  • Prevents the building of deposits
  • Optimises the cleaning results after neutral cleaning
  • Efficient neutralisation after alkaline cleaning
Thermosept® NKZ is used for the reprocessing of surgical and microsurgical instruments, stainless steel containers, rigid endoscopes, anaesthetics material, operating theatre shoes and baby bottles or laboratory equipment after alkaline cleaning, to avoidMore Product Information