Stampa…
Surface disinfectant, mikrozid® AF liquid
Fornitore: SCHUELKE MAYR
109160
SCUE109160EA 0 EUR
SCUE109160
Surface disinfectant, mikrozid® AF liquid
Soluzioni detergenti Disinfettanti
Aldehyde-free rapid disinfectant with broad biocidal activity, used for medical devices.
  • Dries rapidly without leaving smears
  • Good wetting properties
  • Ready to use.