Stampa…
Micro filter funnels, DURAN®
Fornitore: DWK Life Sciences
258575104 258575207 258575301 258575404
SCOT258575104EA 51.2 EUR
SCOT258575104 SCOT258575207 SCOT258575301 SCOT258575404
Micro filter funnels, DURAN®
Imbuti Imbuti per filtrazione Imbuti microfiltranti
DURAN®, borosilicate glass 3.3, clear.
  • For filtration of very small volumes
  • Highly resistant to most chemicals such as solvents, acids and alkalis
  • Very good temperature resistance
With integrated sintered glass filter plate.