Stampa…
Cannon-Fenske viscometers for manual measurements
Fornitore: SI Analytics
285403523 285403507 285403515 285403531 285403548 285403564 285403572 285403589 285403597 285403601 285403618
SCOI285403523EA 336 EUR
SCOI285403523 SCOI285403507 SCOI285403515 SCOI285403531 SCOI285403548 SCOI285403564 SCOI285403572 SCOI285403589 SCOI285403597 SCOI285403601 SCOI285403618
Cannon-Fenske viscometers for manual measurements
Viscosimetri Viscometri Capillari
Cannon-Fenske viscometers for manual measurements especially in routine analyses.
Suitable for transparent liquids, foaming liquids and liquid mixture with highly volatile components.