Stampa…
UV ring lights, VisiLED, S80-55UV
Fornitore: SCHOTT AG Lighting and imaging
400255 400225 400200 400266
630-3504EA 944 EUR
630-3504 SCOC400225 630-3180 SCOC400266
UV ring lights, VisiLED, S80-55UV
Luci Luci ad anello
The unique VisiLED UV ring light allows electronic switching between bright field and UV.
  • Segment ring light, consists of 4 UV and 4 white LED segments
  • The only segment ring light in which white-light and UV-LEDs are alternately installed in eight segments
  • LED arrangement allows objects to be examined from the same illumination angle, significantly improving the ability to compare and reproduce resulting images
  • While the homogenous white light displays samples in natural color, UV excitation reveals previously invisible parts of the sample through