Stampa…
Tork Light Duty Cleaning Cloths
Fornitore: Essity
500479
SCAA500479EA 114 EUR
SCAA500479
Tork Light Duty Cleaning Cloths
Panni Panni per pulizia riutilizzabili Panni per usi generali
Multipurpose cleaning cloths for light-duty cleaning tasks. Use these durable cloths wet or dry to clean dust and dirt, and can rinse and reuse them as needed.
  • These disposable cleaning cloths are compatible with most chemical solvents, so no need to change the cleaning products
  • Compatible Top-pak dispenser has one-at-a-time dispensing to help optimise consumption and minimise waste
Tork light-duty cleaning cloths are an economical, multipurpose wiper. Compatible with most chemical solvents, these durable cloths can be used wet or dry for variousMore Product Information