Stampa…
Tork S5 Sensitive Moisturising Mini Hand Lotion, Non-Perfumed
Fornitore: Essity
425202
SCAA425202EA 127 EUR
SCAA425202
Tork S5 Sensitive Moisturising Mini Hand Lotion, Non-Perfumed
Cura della pelle Creme Lozioni per corpo e mani
Non-perfumed lotion, with canola oil, glycerine and panthenol that help maintain the skin’s moisture balance.
  • Fast absorbing for a non-greasy feel and leaves skin feeling soft and nourished
  • Dermatologically tested with skin-friendly pH
  • 96% of ingredients are from natural origin (according to specification and ISO 16128, including water)
  • Perfume free fragrance, premium quality
  • Colour: White
  • Compatible with S5 dispensers
Tork sensitive moisturising hand lotion is ideal for dry skin, leaving it feeling nourished and soft. This non-perfumed lotion has