Stampa…
PFA tongs
Fornitore: SAVILLEX
700-850
SAVI700-850EA 0 EUR
SAVI700-850
PFA tongs
Pinze
Adjustable PFA tongs are moulded from the highest purity PFA resin for ultratrace metals use.
Service temperature range is –200 to +260 °C (–328 to +500 °F).
Ideal for transferring ICP glassware during cleaning. Tongs are 28 cm (11") long with three adjustable width positions.