Stampa…
Membrane filters, type 131
Fornitore: Sartorius
13101-50-AHN
SART13101-50-AHNEA 299 EUR
SART13101-50-AHN
Membrane filters, type 131
Filtri Membrane filtranti
Non sterile membrane filters for colony counting, particle testing, microscopy and sterility testing. The membrane filters are made of cellulose nitrate (cellulose ester).
  • High flow membranes