Stampa…
Permanent lumocolor felt Pens
Fornitore: SARSTEDT
95.953 95.954 95.955 95.956
SARS95.953EA 12.9 EUR
SARS95.953 SARS95.954 SARS95.955 SARS95.956
Permanent lumocolor felt Pens
Penne Pennarelli indelebili
Permanent lumocolour felt pen for plastic tibes and cups.
  • Clean and smear-roof
  • Water-proof
  • Fast drying
Informazioni per l'ordine: 10 pens (same colour) to a cardboard box.