Stampa…
Microtubes, with thread
Fornitore: SARSTEDT
72.694.005 72.730.006 72.692 72.693 72.693.005 72.692.005 72.694.006 72.730.005 72.607 72.608 72.609.001 72.785.005
SARS72.694.005EA 175 EUR
SARS72.694.005 SARS72.730.006 SARS72.692 SARS72.693 SARS72.693.005 SARS72.692.005 SARS72.694.006 SARS72.730.005 SARS72.607 SARS72.608 SARS72.609.001 SARS72.785.005
Microtubes, with thread
Tubi Microprovette
PP, translucent.
  • Safe screw cap with O-ring
  • Sterile or non sterile
  • Autoclavable
For transport, storage and sample processing in the laboratory.