Stampa…
Reagent/centrifuge tubes, round bottom, with or without push cap
Fornitore: SARSTEDT
55.516 55.484.001 55.460 55.465 55.473 55.484 55.461 55.468 55.463 55.459 55.475.001 55.476 55.518 55.510 55.525 55.538 55.466 55.526 55.475 55.517 55.511 55.468.001 55.514 55.522 55.524 55.526.005 55.533 55.535 62.492 55.466.005 55.467 55.468.005 55.471 55.472 55.474 55.475.005 55.476.005 55.481 55.482 55.483 55.484.005 55.495 55.515 55.523 55.525.005 55.528 55.534 55.539 55.598 55.655 55.705 62.476.022 62.493
SARS55.516EA 87.4 EUR
SARS55.516 SARS55.484.001 SARS55.460 212-3359 212-3360 391-3812 SARS55.461 SARS55.468 SARS55.463 SARS55.459 SARS55.475.001 SARS55.476 SARS55.518 SARS55.510 SARS55.525 SARS55.538 SARS55.466 SARS55.526 SARS55.475 SARS55.517 SARS55.511 SARS55.468.001 SARS55.514 SARS55.522 SARS55.524 SARS55.526.005 SARS55.533 SARS55.535 SARS62.492 SARS55.466.005 SARS55.467 SARS55.468.005 SARS55.471 SARS55.472 SARS55.474 SARS55.475.005 SARS55.476.005 SARS55.481 SARS55.482 SARS55.483 SARS55.484.005 SARS55.495 SARS55.515 SARS55.523 SARS55.525.005 SARS55.528 SARS55.534 SARS55.539 SARS55.598 SARS55.655 SARS55.705 SARS62.476.022 SARS62.493
Reagent/centrifuge tubes, round bottom, with or without push cap
Tubi Tubi per centrifuga
PS, transparent or PP, translucent
  • Non sterile or sterile
  • Plain or with graduation and writing space available
  • Available loose in bags or in StackPack
Universal tubes for a wide range of applications.