Stampa…
Removable powder sampling tips, stainless steel
Fornitore: Sampling Systems
TIP-40R TIP-25R TIP-25P
SAMPTIP-40REA 336 EUR
SAMPTIP-40R SAMPTIP-25R SAMPTIP-25P
Removable powder sampling tips, stainless steel
Campionatori Campionatori per Solidi
Removable tips can add additional effectiveness to powder sampling.
  • Optional range of tips for a variety of environments
  • Unscrew to enable thorough cleaning of the sampler
  • Available in varied diameters and styles
This range of sampling tips provide an excellent alternative to standard tips and can be chosen according to the environment in which the sampler is being used. They can all be unscrewed for easy cleaning and come in a variety of diameters and styles.