Stampa…
Square HDPE Bottle
Fornitore: Sampling Systems
8165A-250 8165A-1000 8165A-500
SAMP8165A-250EA 13.2 EUR
SAMP8165A-250 SAMP8165A-1000 SAMP8165A-500
Square HDPE Bottle
Bottiglie Bottiglie in plastica
Square HDPE bottles for use with a range of liquids and powders.
  • Different sizes available
  • Ribbed sides for easy handling
  • Available from stock
  • Fast delivery
This range of HDPE containers are perfect for use with a range of powders and liquids. They have a handy square space-saving shape and incorporate side ridges for easy handling when wet.