Stampa…
Samplers, MultiThief
Fornitore: Sampling Systems
1290K-1000 1290K-2000 1290W-25 1290W-50 1290K-1500 1290W-10 1290W-05 1290W-00 1290W-60
SAMP1290K-1000EA 2550 EUR
SAMP1290K-1000 SAMP1290K-2000 SAMP1290W-25 SAMP1290W-50 SAMP1290K-1500 SAMP1290W-10 SAMP1290W-05 SAMP1290W-00 300-0422
Samplers, MultiThief
Campionatori Campionatori per Solidi
The MultiThief is designed for the multi-level sampling of free flowing powders and granules.
  • Variable sample volumes up to 60 ml
  • Suitable for free fl owing powders and granules
  • Can be quickly and easily changed by removing cells