Stampa…
Inserts for U-D master manual samplers
Fornitore: Sampling Systems
1230T-05S 1230T-10S 1230T-30S 1230T-03S 1230T-15S
SAMP1230T-05SEA 459 EUR
SAMP1230T-05S SAMP1230T-10S SAMP1230T-30S SAMP1230T-03S SAMP1230T-15S
Inserts for U-D master manual samplers
Campionatori Campionatori per Solidi
Stainless steel inserts for U-D Master manual samplers.
  • Made from 316 stainless steel
These inserts are ideal for helping to prevent sampling error. By simply changing the sample inserts, different volumes can be accurately sampled.