Stampa…

Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Fornitore: Rockland Immunochemicals
710-1532
ROCK710-1532EA 396 EUR
ROCK710-1532
Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorpi
This product has been assayed against 1.0 µg of Mouse IgG in a standard capture ELISA using pNPP p-nitrophenyl phosphate code # NPP-10 as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:30,000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR