Stampa…

Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (Peroxidase)

Fornitore: Rockland Immunochemicals
614-4304
ROCK614-4304EA 421 EUR
ROCK614-4304
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (Peroxidase)
Anticorpi
This product has been assayed against 1.0 µg of Swine IgG F(ab')2  in a standard capture ELISA using ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Pig
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR