Stampa…

Anti-IgG F(c) Rabbit Polyclonal Antibody (Peroxidase)

Fornitore: Rockland Immunochemicals
610-4303
ROCK610-4303EA 421 EUR
ROCK610-4303
Anti-IgG F(c) Rabbit Polyclonal Antibody (Peroxidase)
Anticorpi
Secondary Rabbit Anti-IgG F(c) Reacts with Mouse

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: F(c)
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR