Stampa…

Anti-IgG F(ab')2 Goat Polyclonal Antibody

Fornitore: Rockland Immunochemicals
602-1104
ROCK602-1104EA 248 EUR
ROCK602-1104
Anti-IgG F(ab')2 Goat Polyclonal Antibody
Anticorpi
Secondary Goat Anti-IgG F(ab')2 Reacts with Cat (Feline)

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Cat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR