Stampa…

Anti-IgG (H+L) Rabbit Polyclonal Antibody (Peroxidase)

Fornitore: Rockland Immunochemicals
209-4302
ROCK209-4302EA 617 EUR
ROCK209-4302
Anti-IgG (H+L) Rabbit Polyclonal Antibody (Peroxidase)
Anticorpi
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Peroxidase
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR