Stampa…

Anti-IgG F(ab')2 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fornitore: Rockland Immunochemicals
209-1304
ROCK209-1304EA 634 EUR
ROCK209-1304
Anti-IgG F(ab')2 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpi
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Peroxidase
Epitope:
Host: Goat
Isotype: none
Reactivity: Pan
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR